Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spata18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178387.3 | NP_848474.2 | 11, 12 | 1770 | ||
GenBank mRNA | AK006916.1 | - | 10, 11 | 1449 | ||
BC120503.1 | - | 8, 9 | 1222 | |||
BC120505.1 | - | 8, 9 | 1222 | |||
BC144721.1 | - | 8, 9 | 1222 |