Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam53a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178390.3 | NP_848477.2 | 6 | 1981 | ||
GenBank mRNA | AK010537.1 | - | 6 | 1917 | ||
AK017321.1 | - | 5 | 2326 | |||
AK155077.1 | - | 7 | 2094 | |||
AK163326.1 | - | 6 | 1953 | |||
AK179669.1 | - | 1 | 382 | |||
AK195761.1 | - | 1 | 368 | |||
AK220276.1 | - | 4 | 8200 | |||
BC016596.1 | - | 4 | 1406 | 3257498 | ||
BC028656.1 | - | 6 | 1701 | 1244982 |