Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wipi2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178398.4 | NP_848485.1 | 5 | 684 | ||
GenBank mRNA | AK010881.1 | - | 5 | 712 | ||
AK043857.1 | - | 4 | 1493 | |||
AK155113.1 | - | 5 | 714 | |||
AK169051.1 | - | 5 | 658 | |||
AK172030.1 | - | 5 | 660 | |||
BC044894.1 | - | 5 | 709 | 4235237 |