Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agbl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289656.1 | NP_001276585.1 | 4, 5 | 995 | ||
NM_001289657.1 | NP_001276586.1 | 4, 5 | 995 | |||
NM_178630.4 | NP_848745.2 | 4, 5 | 995 | |||
GenBank mRNA | AK018326.1 | - | 5, 6 | 842 | ||
AK029793.1 | - | 4, 5 | 996 | |||
AK029798.1 | - | 4, 5 | 579 | |||
AK030378.1 | - | 4, 5 | 553 | |||
AK047649.1 | - | 4, 5 | 552 | |||
AK054066.1 | - | 3, 4 | 2026 | |||
BC139065.1 | - | 4, 5 | 447 | 8862087 | ||
DQ867031.1 | - | 4, 5 | 996 | |||
DQ867032.1 | - | 4, 5 | 996 |