Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agbl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289656.1 | NP_001276585.1 | 4 | 909 | ||
NM_001289657.1 | NP_001276586.1 | 4 | 909 | |||
NM_001289658.1 | NP_001276587.1 | 4 | 909 | |||
NM_178630.4 | NP_848745.2 | 4 | 909 | |||
GenBank mRNA | AK013677.1 | - | 1 | 1477 | ||
AK018326.1 | - | 5 | 757 | |||
AK029793.1 | - | 4 | 910 | |||
AK029798.1 | - | 4 | 493 | |||
AK030378.1 | - | 4 | 467 | |||
AK047649.1 | - | 4 | 466 | |||
AK054066.1 | - | 3 | 1940 | |||
BC028521.1 | - | 4 | 421 | |||
BC087897.1 | - | 4 | 452 | |||
BC139065.1 | - | 4 | 361 | 8862087 | ||
DQ867031.1 | - | 4 | 910 | |||
DQ867032.1 | - | 4 | 910 |