Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Optn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181848.4 | NP_862896.1 | 2 | 121 | ||
GenBank mRNA | AY071834.1 | - | 2 | 228 | ||
BC061185.1 | - | 2 | 121 | 30314331 |