Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MCHR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005297.3 | NP_005288.3 | 2 | 1518 | ||
GenBank mRNA | AB063174.1 | - | 2 | 1256 | ||
AF490537.2 | - | 2 | 1601 | |||
AK308739.1 | - | 2 | 1071 | |||
AK314857.1 | - | 2 | 1705 | |||
AY562945.1 | - | 2 | 1250 | |||
AY745811.1 | - | 2 | 1093 | |||
AY747629.1 | - | 3 | 1045 | |||
BC001736.1 | - | 2 | 1463 | 3503484 | ||
BC021146.1 | - | 2 | 1463 | 4890530 | ||
BT006725.1 | - | 2 | 1250 | |||
CR456497.1 | - | 2 | 1306 |
Set Membership: |
G-protein coupled receptor |