Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4930563M21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183111.2 | NP_898934.2 | 7 | 480 | ||
GenBank mRNA | AK016210.1 | - | 7 | 480 |