Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc125 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168386.2 | NP_001161858.1 | 9 | 1226 | ||
NM_183115.5 | NP_898938.4 | 10 | 1304 | |||
GenBank mRNA | AK017973.1 | - | 9 | 989 | ||
BC085249.1 | - | 10 | 1022 | 30726150 |