Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1110008P14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198001.2 | NP_932118.1 | 2 | 575 | ||
GenBank mRNA | AK003598.1 | - | 2 | 530 | ||
AK079725.1 | - | 2 | 495 | |||
AK081091.1 | - | 533 | ||||
BC024615.1 | - | 2 | 542 | 2812475 | ||
BC049639.1 | - | 2 | 494 | 6590746 |