Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164583.1 | NP_001158055.1 | 5 | 650 | ||
NM_001164584.1 | NP_001158056.1 | 5 | 595 | |||
NM_001164585.1 | NP_001158057.1 | 6 | 660 | |||
NM_198412.2 | NP_940804.1 | 6 | 569 | |||
GenBank mRNA | AK122293.1 | - | 5 | 571 | ||
AK147570.1 | - | 6 | 532 | |||
AK147637.1 | - | 6 | 532 | |||
AK163657.1 | - | 5 | 469 | |||
BC060734.1 | - | 5 | 650 | 6409942 | ||
BC064460.1 | - | 6 | 538 | 6821822 |