Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Paqr9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198414.2 | NP_940806.2 | 1 | 1627 | ||
GenBank mRNA | AY424298.1 | - | 1 | 1627 |
Set Membership: |
G-protein coupled receptor |