Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam160a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242363.2 | NP_001229292.1 | 10 | 2102 | ||
NM_001242364.2 | NP_001229293.1 | 9 | 2053 | |||
NM_199009.3 | NP_950174.1 | 9 | 2011 | |||
GenBank mRNA | AK085350.1 | - | 9 | 2007 | ||
AK122552.1 | - | 9 | 1939 | |||
AK154173.1 | - | 10 | 2046 | |||
AK155273.1 | - | 10 | 2047 | |||
AK169996.1 | - | 10 | 2084 | |||
AK216018.1 | - | 2 | 306 | |||
BC018464.1 | - | 9 | 1970 | |||
BC021532.1 | - | 7 | 1380 | |||
BC043717.1 | - | 8 | 1527 | |||
BC063093.1 | - | 9 | 1968 | 6332038 |