Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc30a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022885.2 | NP_075023.2 | 10 | 1493 | ||
GenBank mRNA | AF233321.1 | - | 3 | 512 | ||
AF461761.1 | - | 10 | 1392 | |||
AK007424.1 | - | 1 | 13 | |||
AK051183.1 | - | 10 | 1493 | |||
AK088004.1 | - | 9 | 1237 | |||
AK090176.1 | - | 10 | 1402 | |||
AK166006.1 | - | 10 | 1303 | |||
AK183613.1 | - | 1 | 129 | |||
AK189328.1 | - | 1 | 129 | |||
BC029033.1 | - | 7 | 935 | |||
BC033452.1 | - | 10 | 1392 | 3156622 |