Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc30a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022885.2 | NP_075023.2 | 15 | 2259 | ||
GenBank mRNA | AF233321.1 | - | 8 | 1278 | ||
AF461761.1 | - | 15 | 2158 | |||
AK051183.1 | - | 15 | 2259 | |||
AK088004.1 | - | 14 | 2003 | |||
AK090176.1 | - | 15 | 2168 | |||
AK166006.1 | - | 15 | 2069 | |||
AK215365.1 | - | 2 | 215 | |||
BC029033.1 | - | 12 | 1701 | |||
BC029217.1 | - | 8 | 1111 | |||
BC033452.1 | - | 15 | 2158 | 3156622 |