Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agtpbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001048008.2 | NP_001041473.1 | 10 | 918 | ||
NM_001284218.1 | NP_001271147.1 | 10 | 1043 | |||
NM_001284219.1 | NP_001271148.1 | 9 | 989 | |||
NM_001284221.1 | NP_001271150.1 | 9 | 793 | |||
NM_023328.3 | NP_075817.2 | 10 | 918 | |||
GenBank mRNA | AF219141.1 | - | 10 | 857 | ||
AK015394.1 | - | 9 | 757 | |||
AK051467.1 | - | 9 | 829 | |||
AK132695.1 | - | 10 | 858 | |||
AK133215.1 | - | 10 | 977 | |||
AK170046.1 | - | 10 | 861 | |||
BC042475.1 | - | 9 | 989 | |||
BC060633.1 | - | 10 | 962 | |||
BC082335.1 | - | 9 | 781 | 30536788 | ||
CN527234.1 | - | 7 | 604 |