Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Samsn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023380.2 | NP_075869.2 | 4 | 389 | ||
GenBank mRNA | AF222928.1 | - | 4 | 345 | ||
AK016072.1 | - | 1120 | ||||
AK041340.1 | - | 4 | 321 | |||
AK053946.1 | - | 4 | 364 | |||
BC052906.1 | - | 5 | 401 | 30078141 | ||
BC064778.1 | - | 4 | 389 | 30077237 | ||
DQ333189.1 | - | 4 | 389 |