Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Samsn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023380.2 | NP_075869.2 | 4 | 452 | ||
GenBank mRNA | AF222928.1 | - | 4 | 408 | ||
AK016072.1 | - | 1183 | ||||
AK041340.1 | - | 4 | 384 | |||
AK053946.1 | - | 4 | 427 | |||
BC052906.1 | - | 5 | 464 | 30078141 | ||
BC064778.1 | - | 4 | 452 | 30077237 | ||
DQ333189.1 | - | 4 | 452 |