Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rprm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023396.5 | NP_075885.1 | 1 | 1363 | ||
GenBank mRNA | AK010465.1 | - | 1 | 1362 | ||
BC030065.1 | - | 1 | 1186 | 1434823 |