Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mdp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023397.4 | NP_075886.1 | 2, 3 | 240 | ||
GenBank mRNA | AF230273.1 | - | 2, 3 | 106 | ||
AK007319.1 | - | 2, 3 | 224 | |||
AK007694.1 | - | 2 | 253 | |||
AK008881.1 | - | 2 | 224 | |||
AK160438.1 | - | 2, 3 | 170 | |||
BC046613.1 | - | 2, 3 | 163 | 6465642 | ||
CJ176407.1 | - | 2, 3 | 241 |