Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col4a3bp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164222.1 | NP_001157694.1 | 7 | 1140 | ||
NM_023420.2 | NP_075909.1 | 7 | 1140 | |||
GenBank mRNA | AF232932.1 | - | 7 | 1154 | ||
AF232934.1 | - | 7 | 1154 | |||
AK012989.1 | - | 7 | 1121 | |||
AK030305.1 | - | 8 | 1048 | |||
AK038803.1 | - | 7 | 1121 | |||
AK047100.1 | - | 7 | 1137 | |||
AK080958.1 | - | 7 | 1138 | |||
AK163624.1 | - | 7 | 1130 | |||
BC016197.1 | - | 7 | 1103 | 4502118 |