Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mum1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023431.5 | NP_075920.4 | 5 | 642 | ||
GenBank mRNA | AK078792.1 | - | 2 | 349 | ||
AK154610.1 | - | 5 | 561 | |||
AK158750.1 | - | 5 | 642 | |||
AK159972.1 | - | 4 | 1544 | |||
AK170459.1 | - | 5 | 561 | |||
AK210750.1 | - | 1 | 207 | |||
BC008550.1 | - | 2 | 318 | |||
BC075617.1 | - | 4 | 531 | 5718202 |