Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mum1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023431.5 | NP_075920.4 | 7 | 1310 | ||
GenBank mRNA | AK078792.1 | - | 4 | 1017 | ||
AK147490.1 | - | 2 | 148 | |||
AK154610.1 | - | 7 | 1229 | |||
AK158750.1 | - | 7 | 1310 | |||
AK159972.1 | - | 6 | 2212 | |||
AK170459.1 | - | 7 | 1229 | |||
AK178665.1 | - | 3 | 140 | |||
AK192032.1 | - | 2 | 53 | |||
BC008550.1 | - | 4 | 983 | |||
BC023031.1 | - | 2 | 68 | |||
BC057192.1 | - | 2 | 82 | |||
BC075617.1 | - | 6 | 1199 | 5718202 |