Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serhl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023475.3 | NP_075964.1 | 3, 4 | 415 | ||
GenBank mRNA | AJ245737.1 | - | 418 | |||
AJ251200.1 | - | 3, 4 | 418 | |||
AK002313.1 | - | 3, 4 | 328 | |||
AK003827.1 | - | 3, 4 | 328 | |||
AK154770.1 | - | 2, 3 | 1623 | |||
BC024048.1 | - | 3, 4 | 296 | |||
BC046342.1 | - | 3, 4 | 300 | 6475904 | ||
BC055431.1 | - | 3, 4 | 427 | 3673247 | ||
BC064069.1 | - | 2, 3 | 485 |