Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fahd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023480.2 | NP_075969.1 | 1 | 1051 | ||
GenBank mRNA | AB041600.1 | - | 1 | 885 | ||
AK003914.1 | - | 1 | 841 | |||
AK044920.1 | - | 1 | 850 | |||
AK141973.1 | - | 1 | 1051 | |||
AK157292.1 | - | 1 | 1050 | |||
BC026949.1 | - | 1 | 1037 | 4221757 |