Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNLY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001302758.1 | NP_001289687.1 | 3, 4 | 373 | ||
NM_006433.4 | NP_006424.2 | 2, 3 | 292 | |||
NM_012483.3 | NP_036615.2 | 3, 4 | 536 | |||
XM_005264084.2 | XP_005264141.1 | 3, 4 | 306 | |||
XM_005264085.2 | XP_005264142.1 | 2, 3 | 281 | |||
XM_005264087.2 | XP_005264144.1 | 3, 4 | 533 | |||
GenBank mRNA | AK304728.1 | - | 3, 4 | 306 | ||
AK308781.1 | - | 2, 3 | 395 | |||
AK310057.1 | - | 3, 4 | 534 | |||
AK311537.1 | - | 3, 4 | 426 | |||
AK311538.1 | - | 2, 3 | 159 | |||
AK311657.1 | - | 2, 3 | 822 | |||
BC023576.2 | - | 2, 3 | 167 | 4766109 | ||
CR541859.1 | - | 2, 3 | 164 | |||
DA932688.1 | - | 3, 4 | 534 | |||
X05044.1 | - | 3, 4 | 396 | |||
X54101.1 | - | 2, 3 | 292 |