Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrto4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290810.1 | NP_001277739.1 | 3, 4 | 476 | ||
NM_023536.3 | NP_076025.1 | 3, 4 | 476 | |||
GenBank mRNA | AK011387.1 | - | 3, 4 | 198 | ||
BC005734.1 | - | 3, 4 | 198 | 3598883 | ||
BC096556.1 | - | 3, 4 | 220 | |||
BC138221.1 | - | 3, 4 | 275 | 8861240 | ||
BC145205.1 | - | 3, 4 | 275 | 9053756 | ||
BC145209.1 | - | 3, 4 | 297 |