Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jam2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023844.5 | NP_076333.3 | 5 | 935 | ||
GenBank mRNA | AF255911.1 | - | 5 | 658 | ||
AJ291757.1 | - | 5 | 871 | |||
AK010616.1 | - | 5 | 777 | |||
AK013914.1 | - | 5 | 776 | |||
AK028757.1 | - | 5 | 626 | |||
AK078128.1 | - | 5 | 661 | |||
AK153636.1 | - | 5 | 626 | |||
BC028778.1 | - | 5 | 935 | 1195543 |