Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jam2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023844.5 | NP_076333.3 | 10 | 1576 | ||
GenBank mRNA | AK028757.1 | - | 10 | 1267 | ||
AK153636.1 | - | 10 | 1267 |