Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cep70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023873.3 | NP_076362.2 | 4 | 277 | ||
GenBank mRNA | AF242319.1 | - | 4 | 313 | ||
AK020174.1 | - | 4 | 615 | |||
AK020622.1 | - | 4 | 354 | |||
AK021095.1 | - | 4 | 408 | |||
AK049326.1 | - | 4 | 285 | |||
AK133048.1 | - | 4 | 283 | |||
BC050819.1 | - | 4 | 269 | 6517145 | ||
BC071260.1 | - | 4 | 277 | 6336351 |