Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024196.3 | NP_077158.1 | 2 | 261 | ||
GenBank mRNA | AK006877.1 | - | 2 | 258 | ||
AK036751.1 | - | 171 | ||||
AK076124.1 | - | 2 | 262 | |||
AK082369.1 | - | 257 | ||||
AK085702.1 | - | 258 | ||||
AK158274.1 | - | 258 | ||||
AK168985.1 | - | 262 | ||||
AK172349.1 | - | 258 | ||||
AK188394.1 | - | 1 | 111 | |||
AK212248.1 | - | 2 | 111 | |||
AK218101.1 | - | 2 | 111 | |||
BC002196.1 | - | 1 | 77 | |||
BC040089.1 | - | 2 | 224 | 3991310 | ||
BC086617.1 | - | 248 |