Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mfn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024200.4 | NP_077162.2 | 4 | 582 | ||
GenBank mRNA | AK018181.1 | - | 4 | 579 | ||
AK046313.1 | - | 4 | 535 | |||
AK141611.1 | - | 4 | 583 | |||
AK220169.1 | - | 4 | 570 | |||
AY174062.1 | - | 3 | 711 | |||
BC056641.1 | - | 4 | 541 | 5097345 |