Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mfn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024200.4 | NP_077162.2 | 14 | 1685 | ||
GenBank mRNA | AK018181.1 | - | 14 | 1682 | ||
AK081358.1 | - | 7 | 671 | |||
AK141611.1 | - | 14 | 1686 | |||
AK220169.1 | - | 14 | 1673 | |||
AY174062.1 | - | 13 | 1814 | |||
BC008137.1 | - | 10 | 952 | |||
BC047050.1 | - | 5 | 420 | |||
BC056641.1 | - | 14 | 1644 | 5097345 |