Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Echdc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024208.4 | NP_077170.2 | 5 | 1253 | ||
GenBank mRNA | AK004218.1 | - | 5 | 1255 | ||
AK009166.1 | - | 5 | 1254 | |||
AK209830.1 | - | 1 | 314 | |||
BC002214.1 | - | 5 | 1233 | 3490443 | ||
BC054365.1 | - | 5 | 1222 | 5343451 |