Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042550.1 | NP_001036015.1 | 2, 3 | 526 | ||
NM_001042551.1 | NP_001036016.1 | 2, 3 | 464 | |||
NM_001265602.1 | NP_001252531.1 | 2, 3 | 480 | |||
NM_006444.2 | NP_006435.2 | 2, 3 | 455 | |||
XM_006716933.3 | XP_006716996.1 | 2, 3 | 377 | |||
XM_011518148.2 | XP_011516450.1 | 2, 3 | 340 | |||
XM_011518149.2 | XP_011516451.1 | 2, 3 | 382 | |||
XM_011518152.2 | XP_011516454.1 | 2, 3 | 406 | |||
XM_011518153.1 | XP_011516455.1 | 2, 3 | 406 | |||
XM_017014206.1 | XP_016869695.1 | 2, 3 | 526 | |||
XM_017014207.1 | XP_016869696.1 | 2, 3 | 428 | |||
XM_017014208.1 | XP_016869697.1 | 2, 3 | 468 | |||
GenBank mRNA | AF092563.1 | - | 2, 3 | 390 | ||
AK001485.1 | - | 2, 3 | 512 | |||
AK292599.1 | - | 2, 3 | 403 | |||
AL833191.1 | - | 2, 3 | 464 | |||
AY739716.1 | - | 2, 3 | 356 | |||
BC003396.1 | - | 1, 2 | 199 | |||
BC017845.1 | - | 2, 3 | 394 | |||
BC032705.1 | - | 2, 3 | 513 | |||
BC061906.1 | - | 2, 3 | 412 | |||
BC071750.1 | - | 2, 3 | 526 | |||
BC107891.1 | - | 2, 3 | 500 | |||
BC130385.1 | - | 2, 3 | 310 | 40146416 | ||
BC144163.1 | - | 2, 3 | 326 | 9052683 |