Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024218.4 | NP_077180.1 | 3 | 282 | ||
GenBank mRNA | AK018731.1 | - | 3 | 165 | ||
AK019214.1 | - | 3 | 155 | |||
AK028209.1 | - | 3 | 173 | |||
AK077617.1 | - | 3 | 156 | |||
AK136631.1 | - | 3 | 165 | |||
BC002110.1 | - | 3 | 157 | |||
BC053377.1 | - | 3 | 153 | 4216355 | ||
BC058114.1 | - | 3 | 152 | 6817726 | ||
BC092008.1 | - | 3 | 148 | 30612436 | ||
BY031373.1 | - | 3 | 282 |