Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024218.4 | NP_077180.1 | 6 | 572 | ||
GenBank mRNA | AK018731.1 | - | 6 | 455 | ||
AK077617.1 | - | 6 | 446 | |||
AK136631.1 | - | 6 | 455 | |||
BC002110.1 | - | 6 | 447 | |||
BC053377.1 | - | 6 | 443 | 4216355 | ||
BC058114.1 | - | 6 | 442 | 6817726 | ||
BC092008.1 | - | 6 | 438 | 30612436 |