Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199188.1 | NP_001186117.1 | 11 | 1121 | ||
NM_024225.5 | NP_077187.1 | 10 | 1032 | |||
GenBank mRNA | AK007676.1 | - | 10 | 980 | ||
AK029463.1 | - | 10 | 992 | |||
AK031873.1 | - | 11 | 1013 | |||
AK049129.1 | - | 11 | 1013 | |||
AK078776.1 | - | 11 | 1014 | |||
AK144710.1 | - | 11 | 1017 | |||
AK152311.1 | - | 11 | 1058 | |||
AK159115.1 | - | 7 | 575 | |||
AK159904.1 | - | 10 | 937 | |||
AK167362.1 | - | 10 | 952 | |||
AK169545.1 | - | 9 | 1927 | |||
AK169786.1 | - | 11 | 1013 | |||
AK212372.1 | - | 5 | 347 | |||
AK214918.1 | - | 5 | 372 | |||
BC002242.1 | - | 11 | 1090 | 3492262 |