Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbln7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024237.4 | NP_077199.2 | 7 | 1066 | ||
GenBank mRNA | AK005465.1 | - | 7 | 1018 | ||
BC002308.1 | - | 2 | 142 | |||
BC051550.1 | - | 5 | 622 | |||
BC095941.1 | - | 7 | 998 | 4167912 | ||
BM936626.1 | - | 3 | 327 | |||
EF668007.1 | - | 7 | 922 |