Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rmnd5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024288.2 | NP_077250.2 | 3, 4 | 917 | ||
GenBank mRNA | AK148556.1 | - | 2, 3 | 152 | ||
AK149170.1 | - | 2, 3 | 152 | |||
AK164384.1 | - | 3, 4 | 918 | |||
BC050876.1 | - | 3, 4 | 768 | 6512227 | ||
CF748545.1 | - | 3, 4 | 591 |