Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Derl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024440.2 | NP_077760.1 | 7 | 846 | ||
GenBank mRNA | AB047555.1 | - | 7 | 846 | ||
AK007348.1 | - | 7 | 708 | |||
AK007948.1 | - | 7 | 832 | |||
AK172429.1 | - | 7 | 832 | |||
BC004729.1 | - | 7 | 829 | |||
BC019815.1 | - | 7 | 825 | |||
BC099615.1 | - | 7 | 826 |