Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glod4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026029.3 | NP_080305.2 | 6 | 633 | ||
GenBank mRNA | AK006979.1 | - | 6 | 956 | ||
AK007439.1 | - | 6 | 598 | |||
AK010992.1 | - | 6 | 591 | |||
AK012569.1 | - | 6 | 611 | |||
AK013212.1 | - | 6 | 611 | |||
AK033042.1 | - | 2 | 64 | |||
AK138073.1 | - | 610 | ||||
BC049937.1 | - | 3 | 262 | |||
BC061012.1 | - | 6 | 625 | 6587019 |