Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rrp15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026041.2 | NP_080317.3 | 4 | 619 | ||
GenBank mRNA | AK007616.1 | - | 4 | 573 | ||
AK010658.1 | - | 4 | 587 | |||
AK013217.1 | - | 4 | 587 | |||
AK087997.1 | - | 4 | 589 | |||
AK169088.1 | - | 4 | 620 |