Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rrp15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026041.2 | NP_080317.3 | 5 | 1171 | ||
GenBank mRNA | AK007616.1 | - | 5 | 1125 | ||
AK013204.1 | - | 5 | 1147 | |||
AK087997.1 | - | 5 | 1140 | |||
AK169088.1 | - | 5 | 1172 |