Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prpf18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026045.3 | NP_080321.2 | 10 | 2355 | ||
GenBank mRNA | AK013280.1 | - | 1 | 163 | ||
AK028921.1 | - | 3 | 1492 | |||
AK135707.1 | - | 10 | 2396 | |||
AK135890.1 | - | 10 | 2376 | |||
AK161550.1 | - | 6 | 1799 | |||
BC004573.1 | - | 10 | 2355 | 3586127 | ||
BC019370.1 | - | 1 | 626 | |||
BC028306.1 | - | 10 | 2355 | |||
BC046822.1 | - | 1 | 68 | |||
BC058631.1 | - | 10 | 2350 | 6402639 |
Gene Symbol: Ing4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC082781.1 | - | 105 |