Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp329 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026046.3 | NP_080322.2 | 2 | 1503 | ||
GenBank mRNA | AK013271.1 | - | 2 | 1494 | ||
AK016779.1 | - | 1 | 1320 | |||
AK051535.1 | - | 2 | 1502 | |||
BC072665.1 | - | 2 | 1484 | 30543571 | ||
BC139021.1 | - | 1 | 1473 | 8862043 | ||
BC139023.1 | - | 1 | 1473 | 8862045 |