Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026056.4 | NP_080332.1 | 8 | 1102 | ||
GenBank mRNA | AK010235.1 | - | 8 | 849 | ||
AK013331.1 | - | 8 | 859 | |||
AK014979.1 | - | 4 | 497 | |||
AK033071.1 | - | 8 | 858 | |||
AK035556.1 | - | 8 | 1103 | |||
AK047701.1 | - | 6 | 669 | |||
AK049813.1 | - | 5 | 545 | |||
AK076737.1 | - | 4 | 522 | |||
BC050752.1 | - | 8 | 863 | 6533812 | ||
BC057937.1 | - | 8 | 917 | 5325599 |