Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cers4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026058.4 | NP_080334.3 | 8, 9 | 936 | ||
GenBank mRNA | AK013554.1 | - | 8, 9 | 888 | ||
AK021316.1 | - | 1, 2 | 43 | |||
AK028937.1 | - | 8, 9 | 862 | |||
AK028945.1 | - | 8, 9 | 839 | |||
AK030280.1 | - | 8, 9 | 876 | |||
AK134887.1 | - | 7, 8 | 1595 | |||
AK177826.1 | - | 1, 2 | 62 | |||
AY029531.1 | - | 8, 9 | 837 | |||
BC003946.1 | - | 8, 9 | 823 | 3487343 |