Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dock7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290636.1 | NP_001277565.1 | 42 | 5519 | ||
NM_026082.5 | NP_080358.3 | 41 | 5429 | |||
GenBank mRNA | AK122554.2 | - | 10 | 1157 | ||
AK150619.1 | - | 9 | 1110 | |||
AK152431.1 | - | 9 | 1110 | |||
BC024823.1 | - | 12 | 1531 | |||
BC024917.1 | - | 6 | 684 | |||
BC086672.1 | - | 22 | 2937 | 30647593 | ||
BC094420.1 | - | 29 | 3714 |