Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmco5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026104.4 | NP_080380.1 | 6, 7 | 526 | ||
GenBank mRNA | AK007078.1 | - | 6, 7 | 510 | ||
AK007251.1 | - | 6, 7 | 524 | |||
BC049568.1 | - | 6, 7 | 490 | 6742928 | ||
BU936444.1 | - | 6, 7 | 529 |